Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSCB Monoclonal Antibody (OTI3E1), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA526341
Description
HSCB Monoclonal antibody specifically detects HSCB in Canine, Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
HSCB | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IWL3, Q8K3A0 | |
HSCB | |
Full length human recombinant protein of HSCB produced in HEK293T cell | |
100 μL | |
Primary | |
Canine, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3E1 | |
Unconjugated | |
HSCB | |
AI325508; AW049829; C-HSC20; dJ366L4.2; DnaJ (Hsp40) homolog, subfamily C, member 20; DnaJ homolog subfamily C member 20; DNAJC20; HSC20; HSCB; HscB iron-sulfur cluster co-chaperone homolog; HscB iron-sulfur cluster co-chaperone homolog (E. coli); HscB mitochondrial iron-sulfur cluster cochaperone; HscB mitochondrial iron-sulfur cluster co-chaperone; iron-sulfur cluster co-chaperone protein HscB; Iron-sulfur cluster co-chaperone protein HscB, cytoplasmic; Iron-sulfur cluster co-chaperone protein HscB, mitochondrial; JAC1; J-type co-chaperone HSC20; RGD1311005 | |
Mouse | |
Affinity Chromatography | |
RUO | |
100900, 150274, 360826, 477531 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction