Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSD11B1 Recombinant Rabbit Monoclonal Antibody (JU18-39)

Rabbit Recombinant Monoclonal Antibody

Supplier:  Invitrogen™ MA532813

Catalog No. PIMA532813


Only null left
Add to Cart

Description

Description

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

HSD11B1 is a microsomal enzyme that catalyzes the conversion of the stress hormone cortisol to the inactive metabolite cortisone. In addition, HSD11B1 can catalyze the reverse reaction, the conversion of cortisone to cortisol. Too much cortisol can lead to central obesity, and a particular variation in this gene has been associated with obesity and insulin resistance in children. The protein encoded by this gene is a microsomal enzyme that catalyzes the conversion of the stress hormone cortisol to the inactive metabolite cortisone. In addition, the encoded protein can catalyze the reverse reaction, the conversion of cortisone to cortisol. Too much cortisol can lead to central obesity, and a particular variation in this gene has been associated with obesity and insulin resistance in children. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HSD11B1
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
P16232, P28845, P50172
Hsd11b1
Synthetic peptide within Human HSD11B1 aa 1-50.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
JU18-39
Unconjugated
Hsd11b1
11 beta-hydroxysteroid dehydrogenase type 1; 11beta-HSD type 1; 11-beta-HSD1; 11beta-HSD1; 11beta-HSD-1; 11beta-HSD1A; 11-beta-hydroxysteroid dehydrogenase; 11-beta-hydroxysteroid dehydrogenase 1; 11-beta-hydroxysteroid dehydrogenase type 1; 11beta-hydroxysteroid dehydrogenase type 1; 11-DH; Corticosteroid 11-beta-dehydrogenase isozyme 1; Corticosteroid 11-beta-dehydrogenase, isozyme 1 (11-DH) (11-beta-hydroxysteroid dehydrogenase 1) (11-beta-HSD1) (11beta-HSD1A); cortocosteroid 11-beta dehydrogenase; CORTRD2; HDL; HSD11; HSD11B; Hsd11b1; HSD11L; hydroxysteroid (11-beta) dehydrogenase 1; hydroxysteroid 11-beta dehydrogenase 1; hydroxysteroid dehydrogenase, 11 beta type 1; LRRGT00065; MGC13539; OTTHUMP00000034650; SDR26C1; short chain dehydrogenase/reductase family 26C member 1; short chain dehydrogenase/reductase family 26C, member 1; short-chain reductase; short-chain reductase/dehydrogenase
Rabbit
Protein A
RUO
15483, 25116, 3290
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.