Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD17B12 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP30503320UL

 View more versions of this product

Catalog No. NB096254


Add to cart

Description

Description

HSD17B12 Polyclonal antibody specifically detects HSD17B12 in Human samples. It is validated for Western Blot
Specifications

Specifications

HSD17B12
Unconjugated
Purified
Rabbit
IgG
20 μL
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
PBS with 50% glycerol, pH7.3.
17-beta-HSD 12, 17-beta-hydroxysteroid dehydrogenase 12, EC 1.1.1.62,3-ketoacyl-CoA reductase, EC 1.3.1.-, hydroxysteroid (17-beta) dehydrogenase 12,17beta-HSD type 12, KARestradiol 17-beta-dehydrogenase 12, SDR12C1, short chain dehydrogenase/reductase family 12C, member 1, steroid dehydrogenase homolog
Recombinant fusion protein containing a sequence corresponding to amino acids 43-312 of human HSD17B12 (NP_057226.1).
Affinity purified
RUO
Primary
51144
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit