Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD17B12 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen HSD17B12
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB096253
View Documents
Novus Biologicals
NBP305033100UL
100 μg Each for $320.71
Add to cart
 
NB096254
View Documents
Novus Biologicals
NBP30503320UL
20 μg Each for $138.37
Add to cart
 
Description

Description

HSD17B12 Polyclonal antibody specifically detects HSD17B12 in Human samples. It is validated for Western Blot
Specifications

Specifications

HSD17B12
Unconjugated
Rabbit
Human
17-beta-HSD 12, 17-beta-hydroxysteroid dehydrogenase 12, EC 1.1.1.62,3-ketoacyl-CoA reductase, EC 1.3.1.-, hydroxysteroid (17-beta) dehydrogenase 12,17beta-HSD type 12, KARestradiol 17-beta-dehydrogenase 12, SDR12C1, short chain dehydrogenase/reductase family 12C, member 1, steroid dehydrogenase homolog
Recombinant fusion protein containing a sequence corresponding to amino acids 43-312 of human HSD17B12 (NP_057226.1).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
51144
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit