Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD17B2 Mouse anti-Human, PE/Atto594, Clone: OTI4F10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270945PEATT594

Catalog No. N270945P594


Add to cart

Description

Description

HSD17B2 Monoclonal antibody specifically detects HSD17B2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

HSD17B2
OTI4F10
Flow Cytometry
Affinity Purified, Conjugated
HSD17B2
Full length recombinant protein of human HSD17B2 (NP_002144) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
17-beta-hydroxysteroid dehydrogenase type 2, 20 alpha-hydroxysteroid dehydrogenase, E2DH, EC 1.1.1.62,17-beta-HSD 2, EC 1.1.1.63, EDH17B220-alpha-HSD, estradiol 17-beta-dehydrogenase 2, HSD17, hydroxysteroid (17-beta) dehydrogenase 2, Microsomal 17-beta-hydroxysteroid dehydrogenase, SDR9C2, short chain dehydrogenase/reductase family 9C, member 2, Testosterone 17-beta-dehydrogenase
Mouse
IgG1
0.1 mL
Primary
3294.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit