Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD17B4 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI4C4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270947JF549

Catalog No. N270947J549


Add to cart

Description

Description

HSD17B4 Monoclonal antibody specifically detects HSD17B4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

HSD17B4
OTI4C4
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
HSD17B4
Full length recombinant protein of human HSD17B4 (NP_000405) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
12-alpha-trihydroxy-5-beta-cholest-24-enoyl-CoA hydratase, 7-alpha, D-bifunctional protein, EDH17B4, hydroxysteroid (17-beta) dehydrogenase 4, MPF-2, peroxisomal, peroxisomal multifunctional enzyme type 2,17-beta-HSD IV, peroxisomal multifunctional protein 2,17beta-estradiol dehydrogenase type IV, short chain dehydrogenase/reductase family 8C, member 1,3-alpha
Mouse
IgG1
0.1 mL
Primary
3295.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit