Learn More
HSD17B4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 151161AP
Catalog No. 50-172-9993
Add to cart
Description
Description
Peroxisomal multifunctional enzyme type 2 is a protein that in humans is encoded by the HSD17B4 gene. The protein encoded by this gene is a bifunctional enzyme that is involved in the peroxisomal beta-oxidation pathway for fatty acids. It also acts as a catalyst for the formation of 3-ketoacyl-CoA intermediates from both straight-chain and 2-methyl-branched-chain fatty acids. Defects in this gene that affect the peroxisomal fatty acid beta-oxidation activity are a cause of D-bifunctional protein deficiency (DBPD). An apparent pseudogene of this gene is present on chromosome 8. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
Specifications
HSD17B4
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P51659, P51660, P97852
17 beta HSD 4, D bifunctional protein, DBP, EDH17B4, HSD17B4, MFE 2, SDR8C1
Rabbit
IgG
150 μL
-20°C
Primary
15488, 3295, 79244
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
HSD17B4
Liquid
Hsd17b4
HSD17B4 Fusion Protein Ag7165
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat