Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD17B4 Mouse anti-Human, Clone: OTI2C9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526316

Catalog No. PIMA526316


Add to cart

Description

Description

Bifunctional enzyme acting on the peroxisomal beta-oxidation pathway for fatty acids. Catalyzes the formation of 3-ketoacyl-CoA intermediates from both straight-chain and 2-methyl-branched-chain fatty acids.
Specifications

Specifications

HSD17B4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P51659
DBP, MFE-2, MPF-2, SDR8C1, 17-beta-HSD 4, 17-beta-HSD IV, 17-beta-hydroxysteroid dehydrogenase 4, 17beta-estradiol dehydrogenase type IV, 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholest-24-enoyl-CoA hydratase, D-3-hydroxyacyl-CoA dehydratase, D-bifunctional protein, peroxisomal, beta-hydroxyacyl dehydrogenase, beta-keto-reductase, multifunctional protein 2, peroxisomal multifunctional enzyme type 2, peroxisomal multifunctional protein 2, short chain dehydrogenase/reductase family 8C, member 1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3295
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2C9
Unconjugated
HSD17B4
Liquid
HSD17B4
Full length human recombit protein of HSD17B4 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit