Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSD17B6 Rabbit anti-Human, DyLight 488, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297586G
Description
HSD17B6 Polyclonal specifically detects HSD17B6 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HSD17B6 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
17-beta-HSD 6, 17-beta-HSD6, 17-beta-hydroxysteroid dehydrogenase type 6, 3(alpha->beta)-hydroxysteroid epimerase, 3-alpha->beta-HSE, EC 1.1.1, EC 1.1.1.105, EC 1.1.1.62, EC 1.1.1.63, HSE3-hydroxysteroid epimerase, hydroxysteroid (17-beta) dehydrogenase 6, hydroxysteroid (17-beta) dehydrogenase 6 homolog (mouse), NAD+ -dependent 3 alpha-hydroxysteroid dehydrogenase 3-hydroxysteroid epimerase, Oxidative 3-alpha hydroxysteroid dehydrogenase, oxidative 3-alpha-hydroxysteroid-dehydrogenase, oxidoreductase, retinol dehydrogenase, RODH3(alpha->beta)-hydroxysteroid epimerasel, SDR9C6, short chain dehydrogenase/reductase family 9C, member 6,3-alpha->beta-hydroxysteroid epimerase | |
Produced in rabbits immunized with E. coli-derived Human HSD17B6 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
8630 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title