Learn More
HSD17B6 Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS6592R
Catalog No. 50-198-7360
Add to cart
Description
Description
The protein encoded by this gene has both oxidoreductase and epimerase activities and is involved in androgen catabolism. The oxidoreductase activity can convert 3 alpha-adiol to dihydrotestosterone, while the epimerase activity can convert androsterone to epi-androsterone. Both reactions use NAD+ as the preferred cofactor. This gene is a member of the retinol dehydrogenase family. Transcript variants utilizing alternative polyadenylation signals exist.
Specifications
Specifications
HSD17B6
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HSD17B6
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
8630
HSD17B6
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
O14756
17-beta-HSD 6; 17-beta-HSD6; 17betaHSD9; 17-beta-HSD9; 17-beta-hydroxysteroid dehydrogenase type 6; 3(alpha->beta)-hydroxysteroid epimerase; 3(alpha->beta)-hydroxysteroid epimerasel; 3-alpha->beta-HSE; 3-alpha->beta-hydroxysteroid epimerase; 3-hydroxysteroid epimerase; BOS_5620; Gm182; HSD17B6; Hsd17b9; HSE; hydroxysteroid (17-beta) dehydrogenase 6; hydroxysteroid (17-beta) dehydrogenase 6 homolog; hydroxysteroid (17-beta) dehydrogenase 6 homolog (mouse); hydroxysteroid (17-beta) dehydrogenase 9; hydroxysteroid 17-beta dehydrogenase 6; hydroxysteroid 17-beta dehydrogenase 9; NAD+ -dependent 3 alpha-hydroxysteroid dehydrogenase 3-hydroxysteroid epimerase; oxidative 17 beta hydroxysteroid dehydrogenase type 6; Oxidative 3-alpha hydroxysteroid dehydrogenase; oxidative 3-alpha-hydroxysteroid-dehydrogenase; oxidoreductase; Rdh8; retinol dehydrogenase; RODH; SDR9C6; Short chain dehydrogenase/reductase family 9C member 6; short chain dehydrogenase/reductase family 9C, member 6
KLH conjugated synthetic peptide derived from human HSD17B6.
Protein A
RUO
Antibody
Polyclonal
Human