Learn More
Abnova™ HSD17B7 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00051478T01
Catalog No. 89-947-964
Add to cart
Specifications
Specifications
Western Blot
293T
NM_016371.2
pCMV-HSD17B7 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
HSD17B7 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 37.62kDa
Transient overexpression cell lysate was tested with anti-HSD17B7 antibody (Mfr. No. H00051478-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)