Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD17B7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549584

Catalog No. PA549584

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Responsible for the reduction of the keto group on the C-3 of sterols.
Specifications

Specifications

HSD17B7
Polyclonal
PBS with 0.09% sodium azide
P56937
3-keto-steroid reductase, 17-beta-hydroxysteroid dehydrogenase 7, 17-beta-HSD 7, Estradiol 17-beta-dehydrogenase 7, HSD17B7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51478
Flow Cytometry, Western Blot
Unconjugated
HSD17B7
Liquid
HSD17B7
KLH conjugated synthetic peptide between 57-85 amino acids from the N-terminal region of human HSD17B7.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit