Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSDL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570497
Description
This target displays homology in the following species: Dog: 93%; Human: 100%; Mouse: 79%; Pig: 79%; Rabbit: 93%; Rat: 79%.
HSDL1 gene ontology annotations related to this gene include oxidoreductase activity.
Specifications
HSDL1 | |
Polyclonal | |
Unconjugated | |
Hsdl1 | |
2700067E09Rik; HSDL1; hydroxysteroid dehydrogenase like 1; inactive hydroxysteroid dehydrogenase-like protein 1; RGD1308433; SDR12C3; Short chain dehydrogenase/reductase family 12C member 3; short chain dehydrogenase/reductase family 12C, member 3; steroid dehydrogenase-like protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
83693 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q3SXM5 | |
Hsdl1 | |
synthetic peptide directed towards the N terminal region of human HSDL1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction