Learn More
HSF1 Rat anti-Mouse, DyLight 650, Clone: 10H4, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242205C
Catalog No. NBP242205C
Add to cart
Description
Description
HSF1 Monoclonal antibody specifically detects HSF1 in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
HSF1
10H4
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Purified
HSF1
Purified recombinant mouse HSF1 protein
Protein G purified
Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators
Primary
3297
Mouse
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1
Rat
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Detects approx 85kDa (unstressed cell lysates) and approx 95kDa (heat shocked cell lysates).