Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSF1 Rat anti-Mouse, FITC, Clone: 4B4, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242206F

Catalog No. NBP242206F


Add to cart

Description

Description

HSF1 Monoclonal antibody specifically detects HSF1 in Mouse samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

HSF1
4B4
ELISA, Immunocytochemistry/Immunofluorescence
Purified
HSF1
Purified recombinant mouse HSF1 protein, epitope mapping to amino acids 425-439
Protein G purified
Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators
Primary
3297
Mouse
ELISA, Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1
Rat
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Detects approx 85kDa (unstressed cell lysates) and approx 95kDa (heat shocked cell lysates).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit