Learn More
Invitrogen™ HSF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596200
Description
Positive Samples: 293T, HepG2, SW620, BT-474; Cellular Location: Cytoplasm, Nucleus Immunogen sequence: LDEQRFAKEI LPKYFKHNNM ASFVRQLNMY GFRKVVHIDS GIVKQERDGP VEFQHPYFKQ GQDDLLENIK RKVSSSKPEE NKIRQEDLTK IISSAQKVQI KQETIESRLS ELKSENESLW KEVSELRAKH AQQQQVIRKI VQFIVTLVQN NQLVSLKRKR PLLLNTNGAQ KKNLFQH.
Prokaryotic and eukaryotic cells respond to thermal and chemical stress by inducing a group of genes collectively designated heat shock genes. In eukaryotes, this gene expression is regulated primarily at the transcription level. Heat shock transcription factors (HSF, also designated HSTF) 1 and 2 are involved in this regulation. HSF1 and HSF2 are upregulated by estrogen at both the mRNA and protein level. HSF1 is normally found as a monomer whose transcriptional activity is repressed by constitutive phosphorylation. Upon activation, HSF1 forms trimers, gains DNA binding activity and is translocated to the nucleus. HSF2 activity is associated with differentiation and development, and, like HSF1, binds DNA as a trimer. Both HSF1 and HSF2 are known to be induced by proteasome inhibitors of the ubiquitin pathway.
Specifications
HSF2 | |
Polyclonal | |
Unconjugated | |
HSF2 | |
AI661205; heat shock factor 2; heat shock factor protein 2; Heat shock transcription factor 2; HSF 2; Hsf2; HSTF 2; HSTF2; MGC117376; MGC156196; MGC75048 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3298, 64441 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q03933 | |
HSF2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 39-215 of human HSF2 (Q03933). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.