Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSF2, Rabbit anti-Human, Clone: JE48-40, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP280457
Description
HSF2 Monoclonal antibody specifically detects HSF2 in Human Samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
HSF2 | |
JE48-40 | |
Unconjugated | |
1X TBS(pH7.4), 1%BSA, 50%Glycerol | |
heat shock factor 2, heat shock factor protein 2, heat shock transcription factor 2MGC75048, HSF 2, HSTF 2, HSTF2, MGC117376, MGC156196 | |
Rabbit | |
IgG | |
100 uL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human |
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
Purified | |
HSF2 | |
Synthetic peptide within C-terminal human HSF2. | |
Protein A purified | |
RUO | |
Primary | |
3298 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title