Learn More
HSF2BP Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB30147
Catalog No. 89-033-435
Add to cart
Description
Description
Rabbit polyclonal antibody raised against synthetic peptide of human HSF2BP.
  • High specificity
  • High reactivity
Specifications
Specifications
HSF2BP
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (1.25 ug/mL) The optimal working dilution should be determined by the end user.
NM_007031
HSF2BP
A synthetic peptide corresponding to N-terminus of human HSF2BP.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of human HSF2BP.
In PBS (2% sucrose, 0.09% sodium azide).
Liquid
Rabbit
Protein A purification
RUO
Primary
11077
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)