Learn More
HSF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 228831AP
Catalog No. 50-173-3963
Add to cart
Description
Description
This antibody is specific to human HSF4.
Heat-shock transcription factors (HSFs) activate heat-shock response genes under conditions of heat or other stresses. HSF4 lacks the carboxyl-terminal hydrophobic repeat which is shared among all vertebrate HSFs and has been suggested to be involved in the negative regulation of DNA binding activity. Two alternatively spliced transcripts encoding distinct isoforms and possessing different transcriptional activity have been described.
Specifications
Specifications
HSF4
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9R0L1, Q9ULV5
CTM, Heat shock factor protein 4, hHSF4, HSF 4, HSF4, HSTF 4
Rabbit
IgG
150 μL
-20°C
Primary
26386, 291960, 3299
Western Blot
Unconjugated
HSF4
Liquid
Hsf4
HSF4 Fusion Protein Ag18935
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat