Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSFX1/2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310380100UL
Description
HSFX1/2 Polyclonal specifically detects HSFX1/2 in Human samples. It is validated for Western Blot.Specifications
HSFX1/2 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
HSFX1 heat shock transcription factor family, X linked 1, HSFX2, LW-1 | |
The immunogen is a synthetic peptide directed towards the middle region of human HSFX1/2 (NP_057237). Peptide sequence QVTPQHREPAGPNTQIRSGSAPPATPVMVPDSAVASDNSPVTQPAGEWSE | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
100506164 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction