Learn More
HSFY1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP18012920UL
 View more versions of this product
Catalog No. NBP1802920
Add to cart
Description
Description
HSFY1 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications
Specifications
HSFY1
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
HSFY1
Synthetic peptide directed towards the N terminal of human HSFY1 (NP_149099). Peptide sequence SWDENGTCIVINEELFKKEILETKAPYRIFQTDAIKSFVRQLNLYGFSKI.
45 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 5 ug/ml
Q96LI6
FLJ25453, Heat shock transcription factor 2-like protein, heat shock transcription factor, Y linked 2, heat shock transcription factor, Y-linked, HSF2L, HSF2-like, HSFY, HSFY1
Rabbit
IgG
Protein A purified
RUO
Primary
86614