Learn More
HSG, Heat sink grease, Alfa Aesar™
thermally Conductive Heat Sink Grease Applied To the Base and Mounting Stud of Transistors, Diodes and Rectifiers, As a Coupling Agent
Manufacturer:  Alfa Aesar 445895X
Quantity 4 oz.
 View more versions of this product
Catalog No. AA445895X
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
Paste
Thermal grease
HSG
Specifications
Off-white paste
Odorless
2.3g/cm3
4 oz.
Paste
Thermal grease
Off-white
HSG
Safety and Handling
SDS & Safety Documents
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.