Learn More
Enzo Life Sciences HSP 104 (yeast) polyclonal antibody (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA1040F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of yeast Hsp104. UniProt: P31539. Source: Purified from rabbit serum. Species Reactivity: Yeast. Applications: WB. Recommended dilutions: Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-417