Learn More
Enzo Life SciencesSupplier Diversity Partner HSP 27 polyclonal antibody (200 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADISPA803F
Host: Rabbit. Immunogen: Recombinant human Hsp27. UniProt: P04792. Source: Purified from rabbit serum. Species Reactivity: Human, Porcine, Monkey. Applications: ELISA, IHC, IP, WB. Recommended dilutions: ELISA (1:1,000). Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-536