Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP27 Mouse anti-Human, Mouse (Negative), Rat (Negative), PE-Cyanine7, Clone: 6H11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP225149PECY7

Catalog No. N225149PEC7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

HSP27 Monoclonal antibody specifically detects HSP27 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunoassay.
Specifications

Specifications

HSP27
6H11
Flow Cytometry, Knockout Validated
Affinity Purified, Conjugated
HSPB1
Recombinant full length human HSP27 expressed in and purified from E. coli. [UniProt# P04792]
Immunogen affinity purified
Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific
Primary
3315.0
Flow Cytometry, Immunoassay
PE-Cyanine7
PBS with 0.05% Sodium Azide
28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Mouse (Negative), Rat (Negative)
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit