Learn More
HSP27 Polyclonal Antibody, Invitrogen™
Description
PA1-26358 detects Heat Shock Protein 27 from human and monkey samples. PA1-26358 has been successfully used in immunoprecipitation, immunocyctochemistry, ELISA and Western blot procedures. The PA1-26358 immunogen is recombit human Heat Shock Protein 27 protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Specifications
Specifications
| Antigen | HSP27 |
| Applications | ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Gene | HSPB1 |
| Gene Accession No. | P04792 |
| Gene Alias | Heat shock protein beta-1; HspB1; HSP25; Heat shock 27 kDa protein; HSP 27; Stress-responsive protein 27; SRP27; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein; HSPB1; HSP27 |
| Gene Symbols | HSPB1 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.