Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HSP27 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
Supplier: R&D Systems MAB15801100
This item is not returnable.
View return policy
Description
HSP27 Monoclonal specifically detects HSP27 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
HSP27 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HSPB1, SRP27, Stress-responsive protein 27 | |
E. coli-derived recombinant human HSP27 Met1-Lys205 Accession # P04792 | |
100 μg | |
Primary | |
Detects human HSP27 in direct ELISAs. | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
967060 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P04792 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3315 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?
Provide Content Correction