Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP40/DNAJB1 Mouse anti-Human, Canine, PE-Cyanine7, Clone: OTI1F9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270969PECY7

Catalog No. N270969PEC7


Add to cart

Description

Description

HSP40/DNAJB1 Monoclonal antibody specifically detects HSP40/DNAJB1 in Human, Canine samples. It is validated for Flow Cytometry.
Specifications

Specifications

HSP40/DNAJB1
OTI1F9
Flow Cytometry
Affinity Purified, Conjugated
DNAJB1
Full length recombinant protein of human HSP40/DNAJB1 (NP_006136) produced in HEK293T cells.
Immunogen affinity purified
DNA Repair, Golgi Apparatus Markers, Membrane Trafficking and Chaperones
Primary
3337.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
DnaJ (Hsp40) homolog, subfamily B, member 1, DnaJ (Hsp40) homolog, subfmaily B, member 1, dnaJ homolog subfamily B member 1, DnaJ protein homolog 1, DNAJ1, hDj-1, HDJ1, Heat shock 40 kDa protein 1, heat shock 40kD protein 1, Heat shock protein 40, HSP40, HSPF1Hdj1, Human DnaJ protein 1, radial spoke 16 homolog B, RSPH16B, Sis1
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit