Learn More
Enzo Life SciencesSupplier Diversity Partner HSP 40 Hdj2 (human), (recombinant) (200 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADISPP405F
Alternative name: Heat shock protein 40. DnaJ protein homolog 2. MW: ~54kDa. Purity: ≥90% (SDS-PAGE; Western blot). Formulation: Liquid. In TBS containing 1mM DTT. Source: Produced in E. coli. UniProt: P31689. Application: WB. Recommended dilutions: Western Blot (100ng, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Long Term Storage: -80°C
Catalog No. 50-667-716