Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hsp40/YDJ1 Mouse anti-Yeast, Alexa Fluor™ 350, Clone: 1G10.H8, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212879AF350
Description
Hsp40/YDJ1 Monoclonal specifically detects Hsp40/YDJ1 in Yeast samples. It is validated for Western Blot, ELISA.Specifications
Hsp40/YDJ1 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
YDJ1 | |
Full length protein yeast HSP40 (YDJ1) | |
Primary | |
Detects approx 40kDa. Yeast specific product. | |
Store at 4°C in the dark. |
Western Blot, ELISA | |
1G10.H8 | |
Western Blot, ELISA | |
MAS5, YDJ1 Ydj1p | |
Mouse | |
0.1 mL | |
855661 | |
Yeast | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title