Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp40/YDJ1 Mouse anti-Yeast, Alexa Fluor™ 532, Clone: 1G10.H8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212879AF532

Catalog No. NP212879A32


Add to cart

Description

Description

Hsp40/YDJ1 Monoclonal antibody specifically detects Hsp40/YDJ1 in Yeast samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Hsp40/YDJ1
1G10.H8
Western Blot, ELISA
Conjugated, Purified
YDJ1
Full length protein yeast HSP40 (YDJ1)
Protein G purified
Store at 4°C in the dark.
Monoclonal
Detects approx 40kDa. Yeast specific product.
ELISA, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
MAS5, YDJ1 Ydj1p
Mouse
IgG1 Kappa
0.1 mL
Primary
855661.0
Yeast
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit