Learn More
Enzo Life SciencesSupplier Diversity Partner HSP60 monoclonal antibody (LK-2) (PE conjugate) (50 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADISPA807PED
Clone: LK-2. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human HSP60. UniProt: P10809. Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Ehrlichia sennetsu, Rabbit, Plant, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, C. elegans, Bovine. Applications: Flow Cytometry. Recommended dilutions: Flow Cytometry (1:100). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles. Protect from light.
Catalog No. 50-667-545