Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP60 Mouse anti-Human, Mouse, Canine, Monkey, Alexa Fluor™ 700, Clone: OTI3A2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270967AF700

Catalog No. N270967A700


Add to cart

Description

Description

HSP60 Monoclonal antibody specifically detects HSP60 in Human, Mouse, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

HSP60
OTI3A2
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
HSPD1
Full length recombinant protein of human HSP60 (NP_955472) produced in E.coli.
Immunogen affinity purified
Apoptosis, Cellular Markers, Loading Controls, Membrane Trafficking and Chaperones, Mitochondrial Markers
Primary
3329.0
Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant)
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Canine, Human, Monkey, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit