Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP60 Rabbit anti-Human, Mouse, Allophycocyanin/Cy7, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP177397APCCY7
Description
HSP60 Polyclonal antibody specifically detects HSP60 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
HSP60 | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant) | |
Rabbit | |
IgG | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Polyclonal | |
Human, Mouse |
Flow Cytometry | |
Flow Cytometry | |
Affinity Purified, Conjugated | |
HSPD1 | |
A synthetic peptide made to an internal region of the human Hsp60 protein (within residues 70-150). [Swiss-Prot P10809] | |
Immunogen affinity purified | |
Apoptosis, Cellular Markers, Loading Controls, Membrane Trafficking and Chaperones, Mitochondrial Markers | |
Primary | |
3329.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title