Learn More
HSP60 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 152821AP
Catalog No. 50-173-0118
Add to cart
Description
Description
This gene encodes a member of the chaperonin family. The encoded mitochondrial protein may function as a signaling molecule in the innate immune system. This protein is essential for the folding and assembly of newly imported proteins in the mitochondria. This gene is adjacent to a related family member and the region between the 2 genes functions as a bidirectional promoter. Several pseudogenes have been associated with this gene. Two transcript variants encoding the same protein have been identified for this gene. Mutations associated with this gene cause autosomal recessive spastic paraplegia 13.
Specifications
Specifications
HSP60
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P10809, P63038, P63039
HSP 60, HSP60, HSP65, HSPD1
Rabbit
IgG
150 μL
-20°C
Primary
15510, 3329, 63868
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
HSPD1
Liquid
HSPD1
HSP60 Fusion Protein Ag7401
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat