Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP60 Rabbit anti-Human,Mouse,Rat, Unconjugated, Clone: 1N3, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen HSP60
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB114525MI
View Documents
MilliporeSigma
ZRB114525UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB11454XMI
View Documents
MilliporeSigma
ZRB11454X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

HSP60 Monoclonal antibody specifically detects HSP60 in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

HSP60
Unconjugated
Rabbit
60 kDa heat shock protein mitochondrial;60 kDa chaperonin;Chaperonin 60;CPN60;Heat shock protein 60;HSP-60;HuCHA60;Mitochondrial matrix protein P1;P60 lymphocyte protein
GST/HIs-tagged recombinant fragment corresponding to 65 amino acids from the C-terminal half of human Heat Shock Protein 60, isoform 1.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Mouse, Rat
HSPD1, HSP60
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit