Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP65 (mycobacterial), Mouse anti-Human,Mouse,Rat,Bacteria,Bovine,Drosophila,Guinea Pig,Hamster,Protozoa, Alexa Fluor 700, Clone: 3F7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP197880AF700

Catalog No. NB004721


Add to cart

Description

Description

HSP65 (mycobacterial) Monoclonal specifically detects HSP65 (mycobacterial) in Human, Mouse, Rat, Bacteria, Bovine, Drosophila, Guinea Pig, Hamster, Protozoa samples. It is validated for Western Blot.
Specifications

Specifications

HSP65 (mycobacterial)
3F7
Western Blot
Conjugated
groEL2
Recombinant M. bovis HSP65.
Ammonium sulfate precipitation
RUO
Primary
Human, Mouse, Rat, Bacteria, Bovine, Drosophila, Guinea Pig, Hamster, Protozoa
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
60 kDa chaperonin 2, 65 kDa antigen, Antigen A, Cell wall protein A, GroEL protein 2, groEL-2, groL2, Heat shock protein 65, hsp65, MT0456, MTV037.04, Protein Cpn60-2, Rv0440
Mouse
IgM
0.1 mL
Store at 4°C in the dark.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit