Learn More
Enzo Life SciencesSupplier Diversity Partner HSP 70-A1 (mouse), (recombinant) (50 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADISPP502D
Alternative name: Heat shock protein 70. MW: ~70kDa (observed). Purity: ≥85% (SDS-PAGE; Western blot). Formulation: Liquid. In Dulbecco’s PBS, pH 7.5, containing 5.0mM glutathione. Source: Produced in E. coli. UniProt: Q61696 (Hspa1a), P17879 (Hspa1b). Application: WB, Activity assay. Recommended dilutions: Western Blot (100ng, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Long Term Storage: -80°C
Catalog No. 50-667-717