Learn More
Enzo Life SciencesSupplier Diversity Partner HSP70 High Sensitivity ELISA kit (1x96wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADIEKS715
Only commercially available kit specifically validated and sensitive enough to quantify HSP70 (HSP72) in serum and plasma samples. The HSP70 high sensitivity EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4.5 hours. Absorbance is read at 450 nm. This is the only commercially available kit specifically validated and sensitive enough to quantify HSP70 (HSP72) in serum and plasma samples, as referenced by the Cell Stress Society International. Alternative Name: Heat shock protein 70. HSP72. Sensitivity: 90 pg/ml (range 0.20-12.5 ng/ml). Assay Time: 4.5 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human, Mouse, Rat. Contents: Microtiter plate, Assay buffer 28, Standard, Wash buffer concentrate, Antibody, Conjugate, TMB Substrate, Stop solution 2
Catalog No. 50-659-134