Learn More
Enzo Life SciencesSupplier Diversity Partner HSP 70 HSP 72 monoclonal antibody (C92F3A-5) (AP conjugate) (200 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADISPA810APF
Clone: C92F3A-5. Host: Mouse. Isotype: IgG1. Immunogen: Native human Hsp70. UniProt: P0DMV8 (HSPA1A), P0DMV9 (HSPA1B). Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Teal, Fish, Dog, Chicken, Bovine. Applications: ELISA, WB. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-550