Learn More
Enzo Life SciencesSupplier Diversity Partner HSP 70 HSP 72 (human), (recombinant) (200 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADINSP555F
A molecular chaperone that assists in the folding of emerging polypeptides and the refolding of denatured proteins. Alternative name: Heat shock protein 70. HspA1A. HspA1B. MW: ~72kDa. Endotoxin content: >500EU/mg purified protein (LAL test). Purity: ≥95% (SDS-PAGE; Western blot). Formulation: Liquid. In Dulbecco's PBS. Source: Produced in E. coli. UniProt: P0DMV8 (HSPA1A), P0DMV9 (HSPA1B). Application: ELISA, IHC, WB, in vitro Assay, Apoptosis assay, Antigen microarray, Activity assay. Long Term Storage: -80°C
Catalog No. 50-666-994