Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP70/HSPA1A Mouse anti-Human, Clone: W27, Novus Biologicals™
Mouse Monoclonal Antibody
$176.85 - $344.00
Specifications
Antigen | HSP70/HSPA1A |
---|---|
Clone | W27 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Host Species | Mouse |
Description
HSP70/HSPA1A Monoclonal specifically detects HSP70/HSPA1A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
HSP70/HSPA1A | |
Monoclonal | |
Mouse | |
Cancer, Cellular Markers, Hypoxia, Membrane Trafficking and Chaperones | |
dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B | |
HSPA1A | |
IgG2a | |
Affinity Purified |
W27 | |
Unconjugated | |
RUO | |
Human | |
3303 | |
BALB/C mice were injected with purified HSP70 protein. | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title