Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP70/HSPA1A Rabbit anti-Human, Mouse, Rat, Porcine, Avian, Bovine, Canine, Chicken, Drosophila, Fish, Guinea Pig, Hamster, Plant, Primate, Sheep, Yeast, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
$176.85 - $574.00
Specifications
Antigen | HSP70/HSPA1A |
---|---|
Dilution | Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 10 ug/ml |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19787202
|
Novus Biologicals
NBP1978720.2MG |
0.2 mg |
Each for $574.00
|
|
NBP19787205
|
Novus Biologicals
NBP1978720.05MG |
0.05 mg |
Each for $321.00
|
|
NB018820
|
Novus Biologicals
NBP1978720.025MG |
0.025 mg |
Each for $176.85
|
|
Description
HSP70/HSPA1A Polyclonal specifically detects HSP70/HSPA1A in Human, Mouse, Rat, Porcine, Avian, Bovine, Canine, Chicken, Drosophila, Fish, Guinea Pig, Hamster, Plant, Primate, Sheep, Yeast samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HSP70/HSPA1A | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Drosophila, Fish, Guinea Pig, Hamster, Plant, Primate, Sheep | |
Q07439 | |
3303 | |
Synthetic peptide corresponding to a portion of rat Hsp70. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 10 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS and 50% Glycerol with 0.09% Sodium Azide | |
dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B | |
HSPA1A | |
IgG | |
Protein A purified |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title