Learn More
HSP70/HSPA1A Rat anti-Drosophila, DyLight 550, Clone: 7FB, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259342R
Catalog No. NBP259342R
Add to cart
Description
Description
HSP70/HSPA1A Monoclonal antibody specifically detects HSP70/HSPA1A in Drosophila samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
HSP70/HSPA1A
7FB
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Purified
HSPA1A
Prepared from Drosophila tissue culture cells heat shocked at 36.5C for 3 hours, and isolated using SDS PAGE.
Protein G purified
Cancer, Cellular Markers, Chaperone Mediated Autophagy (CMA), Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific
Primary
3303
Drosophila
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B
Rat
IgG2b
100 ul
Store at 4C in the dark.
Monoclonal
Detects 70kDa (heat-inducible form).