Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP70/HSPA1A Antibody (7FB), FITC, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP259342F
Description
HSP70/HSPA1A Monoclonal specifically detects HSP70/HSPA1A in Drosophila samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
HSP70/HSPA1A | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
HSPA1A | |
Prepared from Drosophila tissue culture cells heat shocked at 36.5C for 3 hours, and isolated using SDS PAGE. | |
100 μL | |
Primary | |
Detects 70kDa (heat-inducible form). | |
Store at 4C in the dark. | |
IgG2b |
ELISA, Immunocytochemistry, Immunofluorescence | |
7FB | |
ELISA, Immunocytochemistry/Immunofluorescence | |
dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B | |
Rat | |
Protein G purified | |
Cancer, Cellular Markers, Chaperone Mediated Autophagy (CMA), Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific | |
3303 | |
Drosophila | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction