Learn More
Enzo Life Sciences HSP 70 (low endotoxin) (human), (recombinant) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADIESP555F
Encompass_Preferred
Low endotoxin HSP70 recombinant protein. Alternative name: Heat shock protein 70. HspA1A. HspA1B. HSP72. MW: ~72kDa. Endotoxin content: <50EU/mg purified protein (LAL test). Purity: ≥90% (SDS-PAGE; Western blot). Formulation: Liquid. In Dulbecco's PBS. Source: Produced in E. coli. UniProt: P0DMV8 (HSPA1A), P0DMV9 (HSPA1B). Application: ELISA, WB, in vitro Assay, Activity assay. Long Term Storage: -80°C
Catalog No. 50-659-149