Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp70 interacting protein HIP Mouse anti-Human, Alexa Fluor™ 350, Clone: 5C6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242671AF350

Catalog No. NP242671A30


Add to cart

Description

Description

Hsp70 interacting protein HIP Monoclonal antibody specifically detects Hsp70 interacting protein HIP in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Hsp70 interacting protein HIP
5C6
Western Blot, ELISA
Conjugated, Purified
ST13
Recombinant human HIP/ST13 (1-369aa) purified from E. coli
Protein A purified
Lipid and Metabolism, Membrane Trafficking and Chaperones
Primary
6767.0
ELISA, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
AAG2, aging-associated protein 2, FAM10A1FAM10A4, heat shock 70kD protein binding protein, Hip, HIPFLJ27260, HOP, hsc70-interacting protein, Hsp70-interacting protein, HSPABP1, P48MGC129952, PRO0786, Progesterone receptor-associated p48 protein, Protein FAM10A1, Putative tumor suppressor ST13, Renal carcinoma antigen NY-REN-33, SNC6HSPABP, suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein), suppression of tumorigenicity 13 (colon carcinoma) (Hsp70-interacting protein), Suppression of tumorigenicity 13 protein
Mouse
IgG2a Kappa
100 μL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit