Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp70interactingproteinHIP Mouse anti-Human, Janelia Fluor 646, Clone: 1F1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP126368JF646

Catalog No. NBP12368646

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Hsp70interactingproteinHIPMonoclonalantibody specifically detects Hsp70interactingproteinHIP in Human samples.
Specifications

Specifications

Hsp70interactingproteinHIP
1F1
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
ST13
Recombinant human HIP/ST13 (1-369aa) purified from E. coli
Protein G purified
Membrane Trafficking and Chaperones
Primary
6767
Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
AAG2, aging-associated protein 2, FAM10A1FAM10A4, heat shock 70kD protein binding protein, Hip, HIPFLJ27260, HOP, hsc70-interacting protein, Hsp70-interacting protein, HSPABP1, P48MGC129952, PRO0786, Progesterone receptor-associated p48 protein, Protein FAM10A1, Putative tumor suppressor ST13, Renal carcinoma antigen NY-REN-33, SNC6HSPABP, suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein), suppression of tumorigenicity 13 (colon carcinoma) (Hsp70-interacting protein), Suppression of tumorigenicity 13 protein
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit