Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

HSP72 Rat anti-Human, Clone: 3G7, Active Motif™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Specifications

Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Antigen HSP72
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3236 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
61210
10 μg
Each for $116.85
N/A
50-199-3239 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
61209
100 μg
Each for $356.95
N/A
Description

Description

This intronless gene encodes a 70kDa heat shock protein which is a member of the heat shock protein 70 family. In conjuction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. It is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins.
Specifications

Specifications

Monoclonal
Primary
HSP72
Unconjugated
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit