Learn More
Enzo Life SciencesSupplier Diversity Partner HSP90 monoclonal antibody (AC88) (200 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADISPA830F
Clone: AC88. Host: Mouse. Isotype: IgG1. Immunogen: Achlya ambisexualis (water mold) Hsp90. UniProt: Q8LLI6. Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Fusobacterium nucleatum, Water mold, Sheep, Scallop, Rabbit, Porcine, Mussel, Monkey, Mycobacterium leprae, Hamster, Guinea pig, Fish, E. coli, Dog, Chicken, C. elegans, Beluga. Applications: IHC (PS), WB. Recommended dilutions: Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-619